siRNA Details
| Gene Symbol: Cyp2c29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007815.3 | NP_031841.3 | 7, 8 | 1187 | ||
| GenBank mRNA | AK139760.1 | - | 1227 | |||
| AK149558.1 | - | 7, 8 | 1006 | |||
| BC019908.1 | - | 7, 8 | 1169 | 5123631 | ||
| D17674.1 | - | 7, 8 | 1161 | |||

Cyp2c29
Genomic Map