siRNA Details
| Gene Symbol: Cyp2c37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010001.2 | NP_034131.2 | 2, 3 | 349 | ||
| GenBank mRNA | AF047542.1 | - | 333 | |||
| AK005017.1 | - | 2, 3 | 350 | |||
| BC057912.1 | - | 334 | 5052873 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2c37| Gene Symbol: Cyp2c37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010001.2 | NP_034131.2 | 2, 3 | 349 | ||
| GenBank mRNA | AF047542.1 | - | 333 | |||
| AK005017.1 | - | 2, 3 | 350 | |||
| BC057912.1 | - | 334 | 5052873 | |||