siRNA Details
| Gene Symbol: Cyp2c39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010003.2 | NP_034133.2 | 5 | 831 | ||
| GenBank mRNA | AF047726.1 | - | 5 | 782 | ||
| BC057911.1 | - | 5 | 792 | |||
| BC139290.1 | - | 5 | 831 | 8862312 | ||
| BC139291.1 | - | 5 | 831 | 8862313 | ||

Cyp2c39
Genomic Map