siRNA Details
| Gene Symbol: Cyp2d10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010005.3 | NP_034135.2 | 9 | 1519 | ||
| GenBank mRNA | AA239803.1 | - | 2 | 216 | ||
| BC010989.1 | - | 9 | 1517 | 4193743 | ||
| BC051039.1 | - | 8 | 1354 | |||
| BC057924.1 | - | 9 | 1506 | 5100459 | ||
| M27167.1 | - | 9 | 1497 | |||

Cyp2d10
Genomic Map