siRNA Details
| Gene Symbol: Cyp4a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010011.3 | NP_034141.3 | 6 | 730 | ||
| GenBank mRNA | AB017785.1 | - | 6 | 690 | ||
| AB018421.1 | - | 6 | 724 | |||
| AK002528.1 | - | 6 | 731 | |||
| AK098101.1 | - | 6 | 728 | |||
| BC010747.1 | - | 6 | 715 | 4242518 | ||
| BC051049.1 | - | 6 | 716 | 4234581 | ||

Cyp4a10
Genomic Map