siRNA Details
| Gene Symbol: Cyp7b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007825.4 | NP_031851.3 | 6 | 1594 | ||
| GenBank mRNA | AK013034.1 | - | 6 | 1524 | ||
| AK140100.1 | - | 6 | 1586 | |||
| AK172152.1 | - | 6 | 1595 | |||
| BC038810.1 | - | 6 | 1554 | 5098739 | ||
| U36993.1 | - | 6 | 1539 | |||

Cyp7b1
Genomic Map