siRNA Details
| Gene Symbol: Dao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286396.1 | NP_001273325.1 | 10, 11 | 959 | ||
| NM_001286397.1 | NP_001273326.1 | 9, 10 | 851 | |||
| NM_010018.3 | NP_034148.2 | 10, 11 | 1118 | |||
| GenBank mRNA | AK134813.1 | - | 10, 11 | 979 | ||
| BC015269.1 | - | 9, 10 | 851 | 4239778 | ||
| BC018377.1 | - | 10, 11 | 959 | 4242375 | ||
| D10210.1 | - | 9, 10 | 913 | |||
| D10211.1 | - | 9, 10 | 910 | |||
| M32299.1 | - | 10, 11 | 975 | |||

Dao
Genomic Map