siRNA Details
| Gene Symbol: Dbh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138942.3 | NP_620392.2 | 1 | 195 | ||
| GenBank mRNA | AK132245.1 | - | 1 | 196 | ||
| BC141022.1 | - | 1 | 222 | 9055634 | ||
| BC171949.1 | - | 1 | 222 | 9054235 | ||
| S50200.1 | - | 1 | 470 | |||

Dbh
Genomic Map