siRNA Details
| Gene Symbol: Dbnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146308.1 | NP_001139780.1 | 1, 2 | 165 | ||
| NM_001146309.1 | NP_001139781.1 | 1, 2 | 165 | |||
| NM_013810.3 | NP_038838.1 | 1, 2 | 165 | |||
| GenBank mRNA | AK046073.1 | - | 1, 2 | 145 | ||
| AK053796.1 | - | 1, 2 | 161 | |||
| AK078082.1 | - | 1, 2 | 132 | |||
| AK134136.1 | - | 1, 2 | 132 | |||
| AK134487.1 | - | 1, 2 | 132 | |||
| AK140752.1 | - | 1, 2 | 132 | |||
| AK142818.1 | - | 1, 2 | 115 | |||
| AK146920.1 | - | 1, 2 | 132 | |||
| AK151096.1 | - | 1, 2 | 145 | |||
| AK152110.1 | - | 1, 2 | 132 | |||
| AK152204.1 | - | 1, 2 | 145 | |||
| AK153072.1 | - | 1, 2 | 145 | |||
| AK153389.1 | - | 1, 2 | 119 | |||
| AK159984.1 | - | 1, 2 | 142 | |||
| AK161973.1 | - | 1, 2 | 145 | |||
| AK167956.1 | - | 1, 2 | 105 | |||
| AK172471.1 | - | 1, 2 | 122 | |||
| AK199448.1 | - | 1, 2 | 42 | |||
| AY098595.1 | - | 1, 2 | 88 | |||
| BC046430.1 | - | 1, 2 | 130 | 4986658 | ||
| BY293844.1 | - | 1, 2 | 165 | |||
| U58884.1 | - | 1, 2 | 118 | |||

Dbnl
Genomic Map