siRNA Details
| Gene Symbol: Dbnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146308.1 | NP_001139780.1 | 10 | 973 | ||
| NM_001146309.1 | NP_001139781.1 | 10 | 961 | |||
| NM_013810.3 | NP_038838.1 | 10 | 964 | |||
| GenBank mRNA | AK046073.1 | - | 10 | 941 | ||
| AK053796.1 | - | 10 | 957 | |||
| AK078082.1 | - | 10 | 928 | |||
| AK134136.1 | - | 10 | 928 | |||
| AK134487.1 | - | 10 | 940 | |||
| AK140752.1 | - | 10 | 931 | |||
| AK142818.1 | - | 10 | 911 | |||
| AK146920.1 | - | 10 | 928 | |||
| AK151096.1 | - | 10 | 941 | |||
| AK152110.1 | - | 10 | 929 | |||
| AK152204.1 | - | 10 | 941 | |||
| AK153072.1 | - | 10 | 940 | |||
| AK153389.1 | - | 10 | 918 | |||
| AK159984.1 | - | 10 | 938 | |||
| AK161973.1 | - | 9 | 1235 | |||
| AK167956.1 | - | 10 | 900 | |||
| AK168898.1 | - | 10 | 938 | |||
| AK172471.1 | - | 10 | 918 | |||
| AK191372.1 | - | 1 | 40 | |||
| AK198437.1 | - | 1 | 40 | |||
| AY098595.1 | - | 10 | 896 | |||
| BC046430.1 | - | 10 | 926 | 4986658 | ||
| U58884.1 | - | 10 | 917 | |||

Dbnl
Genomic Map