siRNA Details
| Gene Symbol: Dbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010022.3 | NP_034152.2 | 9, 10 | 1275 | ||
| GenBank mRNA | AK075831.1 | - | 9, 10 | 1257 | ||
| AK165959.1 | - | 9, 10 | 1285 | |||
| BC055890.1 | - | 9, 10 | 1231 | 4242586 | ||
| L42996.1 | - | 9, 10 | 1228 | |||

Dbt
Genomic Map