siRNA Details
| Gene Symbol: Dbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010022.3 | NP_034152.2 | 2 | 191 | ||
| GenBank mRNA | AK075831.1 | - | 2 | 171 | ||
| AK165959.1 | - | 2 | 201 | |||
| BC055890.1 | - | 2 | 147 | 4242586 | ||
| L42996.1 | - | 2 | 144 | |||

Dbt
Genomic Map