siRNA Details
| Gene Symbol: Dbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005232.1 | NP_001005232.1 | 2 | 558 | ||
| GenBank mRNA | BC082541.1 | - | 2 | 558 | 5718470 | |
| S75837.1 | - | 2 | 571 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dbx1| Gene Symbol: Dbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005232.1 | NP_001005232.1 | 2 | 558 | ||
| GenBank mRNA | BC082541.1 | - | 2 | 558 | 5718470 | |
| S75837.1 | - | 2 | 571 | |||