siRNA Details
| Gene Symbol: Eci1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010023.4 | NP_034153.2 | 6 | 561 | ||
| GenBank mRNA | AK004838.1 | - | 6 | 562 | ||
| AK029481.1 | - | 6 | 562 | |||
| AK077386.1 | - | 6 | 552 | |||
| BC022712.1 | - | 6 | 545 | 4487371 | ||
| BC054444.1 | - | 6 | 548 | 6476161 | ||
| Z14049.1 | - | 6 | 541 | |||

Eci1
Genomic Map