siRNA Details
| Gene Symbol: Dcpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019910.2 | NP_064294.2 | 3, 4 | 214 | ||
| GenBank mRNA | BC005655.1 | - | 211 | 3709995 | ||
| BY112697.1 | - | 3, 4 | 211 | |||
| BY150555.1 | - | 3, 4 | 214 | |||
| Gene Symbol: Dcpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039238.2 | NP_001034327.2 | 3, 4 | 209 | ||

Dcpp1
Genomic Map