siRNA Details
| Gene Symbol: Dcpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019910.2 | NP_064294.2 | 3 | 190 | ||
| GenBank mRNA | BC005655.1 | - | 187 | 3709995 | ||
| BY112697.1 | - | 3 | 187 | |||
| BY150555.1 | - | 3 | 190 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcpp1| Gene Symbol: Dcpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019910.2 | NP_064294.2 | 3 | 190 | ||
| GenBank mRNA | BC005655.1 | - | 187 | 3709995 | ||
| BY112697.1 | - | 3 | 187 | |||
| BY150555.1 | - | 3 | 190 | |||