siRNA Details
| Gene Symbol: Dctn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198866.1 | NP_001185795.1 | 18 | 2079 | ||
| NM_001198867.1 | NP_001185796.1 | 15 | 2019 | |||
| NM_007835.2 | NP_031861.2 | 18 | 2325 | |||
| GenBank mRNA | AK147769.1 | - | 11 | 1499 | ||
| AK157867.1 | - | 18 | 2326 | |||
| AK171376.1 | - | 15 | 2006 | |||
| AK172261.1 | - | 15 | 1993 | |||
| BC066061.1 | - | 18 | 2052 | 6849060 | ||
| U60312.1 | - | 18 | 2060 | |||

Dctn1
Genomic Map