siRNA Details
| Gene Symbol: Dctn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198866.1 | NP_001185795.1 | 24 | 2889 | ||
| NM_001198867.1 | NP_001185796.1 | 21 | 2829 | |||
| NM_007835.2 | NP_031861.2 | 24 | 3135 | |||
| GenBank mRNA | AK157867.1 | - | 24 | 3136 | ||
| AK171376.1 | - | 21 | 2816 | |||
| AK172261.1 | - | 21 | 2803 | |||
| BC066061.1 | - | 24 | 2862 | 6849060 | ||
| U60312.1 | - | 24 | 2870 | |||

Dctn1
Genomic Map