siRNA Details
| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007838.2 | NP_031864.2 | 4 | 583 | ||
| GenBank mRNA | AK081542.1 | - | 4 | 475 | ||
| AK084267.1 | - | 4 | 467 | |||
| AK150683.1 | - | 4 | 474 | |||
| AK182353.1 | - | 2 | 119 | |||
| AK200087.1 | - | 2 | 144 | |||
| BC068132.1 | - | 4 | 583 | 6808198 | ||
| D89063.1 | - | 4 | 585 | |||

Ddost
Genomic Map