siRNA Details
| Gene Symbol: Ddx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110826.1 | NP_001104296.1 | 5 | 753 | ||
| NM_007841.4 | NP_031867.1 | 5 | 696 | |||
| NM_181324.3 | NP_851841.2 | 5 | 721 | |||
| GenBank mRNA | AF038995.1 | - | 4 | 315 | ||
| AK036648.1 | - | 5 | 749 | |||
| AK054144.1 | - | 5 | 715 | |||
| AK148483.1 | - | 5 | 718 | |||
| BC021452.1 | - | 4 | 613 | 5043599 | ||
| CD353843.1 | - | 5 | 683 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001109292, XM_006242950, XM_006242952, XM_006242954, XM_006242951.

Ddx6
Genomic Map