siRNA Details
| Gene Symbol: Twist2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007855.2 | NP_031881.1 | 2 | 962 | ||
| GenBank mRNA | AK008388.1 | - | 2 | 700 | ||
| AK008411.1 | - | 2 | 699 | |||
| AK011180.1 | - | 2 | 978 | |||
| AK011861.1 | - | 2 | 979 | |||
| BC090636.1 | - | 2 | 967 | 30942245 | ||
| U36384.1 | - | 2 | 962 | |||

Twist2
Genomic Map