siRNA Details
| Gene Symbol: Slc25a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153150.2 | NP_694790.1 | 8 | 936 | ||
| GenBank mRNA | AK005070.1 | - | 8 | 866 | ||
| AK159289.1 | - | 8 | 899 | |||
| AK170191.1 | - | 8 | 900 | |||
| BC037087.1 | - | 8 | 896 | 5124403 | ||

Slc25a1
Genomic Map