siRNA Details
| Gene Symbol: Dhcr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007856.2 | NP_031882.1 | 8 | 1362 | ||
| GenBank mRNA | AF057368.1 | - | 7 | 894 | ||
| AK146811.1 | - | 7 | 885 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhcr7| Gene Symbol: Dhcr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007856.2 | NP_031882.1 | 8 | 1362 | ||
| GenBank mRNA | AF057368.1 | - | 7 | 894 | ||
| AK146811.1 | - | 7 | 885 | |||