siRNA Details
| Gene Symbol: Dhfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010049.3 | NP_034179.1 | 4 | 774 | ||
| GenBank mRNA | AK031241.1 | - | 4 | 408 | ||
| AK136212.1 | - | 4 | 832 | |||
| BC005796.1 | - | 4 | 346 | 3707582 | ||
| L26316.1 | - | 4 | 353 | |||
| M10811.1 | - | 2 | 151 | |||
| V00731.1 | - | 431 | ||||
| V00734.1 | - | 4 | 410 | |||
| X56066.1 | - | 4 | 320 | |||

Dhfr
Genomic Map