siRNA Details
| Gene Symbol: HDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002112.3 | NP_002103.2 | 10, 11 | 1476 | ||
| XM_017022094.1 | XP_016877583.1 | 10, 11 | 1345 | |||
| XM_017022096.1 | XP_016877585.1 | 9, 10 | 1130 | |||
| XM_017022097.1 | XP_016877586.1 | 8, 9 | 1428 | |||
| XM_017022098.1 | XP_016877587.1 | 7, 8 | 948 | |||
| GenBank mRNA | AK027221.1 | - | 3, 4 | 879 | ||
| BC130527.1 | - | 10, 11 | 1177 | 40146558 | ||
| M60445.1 | - | 10, 11 | 1185 | |||
| X54297.1 | - | 10, 11 | 1236 | |||

HDC
Genomic Map