siRNA Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012707.1 | - | 3 | 507 | ||
| AK140734.1 | - | 4 | 427 | |||
| AK220466.1 | - | 4 | 436 | |||
| BC021396.1 | - | 3 | 466 | |||
| BC070412.1 | - | 5 | 601 | 5705112 | ||
| U96963.1 | - | 3 | 439 | |||

Diap1
Genomic Map