siRNA Details
| Gene Symbol: Dio1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007860.3 | NP_031886.3 | 1 | 224 | ||
| GenBank mRNA | AK002549.1 | - | 1 | 225 | ||
| BC019786.1 | - | 1 | 249 | 5133568 | ||
| BC037080.1 | - | 1 | 260 | |||
| BC089608.1 | - | 1 | 224 | 6766076 | ||
| U49861.1 | - | 1 | 224 | |||

Dio1
Genomic Map