siRNA Details
| Gene Symbol: Dio2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010050.2 | NP_034180.1 | 2 | 858 | ||
| GenBank mRNA | AF093137.1 | - | 2 | 716 | ||
| AF096875.1 | - | 2 | 858 | |||
| AF177196.1 | - | 2 | 728 | |||
| AK032038.1 | - | 2 | 568 | |||
| AK045607.1 | - | 2 | 612 | |||
| BC125383.1 | - | 2 | 753 | 40129898 | ||
| BC125385.1 | - | 2 | 753 | 40129900 | ||

Dio2
Genomic Map