siRNA Details
| Gene Symbol: Dll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007866.2 | NP_031892.2 | 5, 6 | 902 | ||
| GenBank mRNA | AB013440.1 | - | 5, 6 | 889 | ||
| AK144282.1 | - | 5, 6 | 903 | |||
| BC052002.1 | - | 5, 6 | 879 | 6404029 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dll3| Gene Symbol: Dll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007866.2 | NP_031892.2 | 5, 6 | 902 | ||
| GenBank mRNA | AB013440.1 | - | 5, 6 | 889 | ||
| AK144282.1 | - | 5, 6 | 903 | |||
| BC052002.1 | - | 5, 6 | 879 | 6404029 | ||