siRNA Details
| Gene Symbol: Dll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007866.2 | NP_031892.2 | 2 | 123 | ||
| GenBank mRNA | AK144282.1 | - | 2 | 124 | ||
| BB868475.1 | - | 2 | 123 | |||
| BC052002.1 | - | 2 | 100 | 6404029 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dll3| Gene Symbol: Dll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007866.2 | NP_031892.2 | 2 | 123 | ||
| GenBank mRNA | AK144282.1 | - | 2 | 124 | ||
| BB868475.1 | - | 2 | 123 | |||
| BC052002.1 | - | 2 | 100 | 6404029 | ||