siRNA Details
| Gene Symbol: Dmwd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010058.2 | NP_034188.2 | 3 | 1865 | ||
| GenBank mRNA | AK086899.1 | - | 2 | 1427 | ||
| BC031193.1 | - | 1 | 1034 | 5054380 | ||
| BC089027.1 | - | 3 | 2097 | 30615314 | ||
| S60312.1 | - | 3 | 1505 | |||

Dmwd
Genomic Map