siRNA Details
| Gene Symbol: Dmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016779.2 | NP_058059.2 | 6 | 1466 | ||
| GenBank mRNA | AK132177.1 | - | 6 | 1467 | ||
| AK157973.1 | - | 6 | 1467 | |||
| BC113753.1 | - | 6 | 1388 | 40039844 | ||
| U65020.1 | - | 6 | 1472 | |||

Dmp1
Genomic Map