siRNA Details
| Gene Symbol: Dnase2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010062.3 | NP_034192.1 | 5 | 741 | ||
| GenBank mRNA | AK145192.1 | - | 5 | 742 | ||
| AK167277.1 | - | 5 | 707 | |||
| BC058609.1 | - | 5 | 688 | 5397016 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnase2a| Gene Symbol: Dnase2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010062.3 | NP_034192.1 | 5 | 741 | ||
| GenBank mRNA | AK145192.1 | - | 5 | 742 | ||
| AK167277.1 | - | 5 | 707 | |||
| BC058609.1 | - | 5 | 688 | 5397016 | ||