siRNA Details
| Gene Symbol: Dnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301737.1 | NP_001288666.1 | 19 | 2116 | ||
| NM_010065.3 | NP_034195.2 | 18 | 2104 | |||
| GenBank mRNA | AK144142.1 | - | 18 | 2055 | ||
| AK220483.1 | - | 19 | 2058 | |||
| BC034679.1 | - | 19 | 2070 | 4506865 | ||
| BC058623.1 | - | 19 | 2050 | 5706803 | ||
| BM950315.1 | - | 5 | 509 | |||
| CO425806.1 | - | 5 | 435 | |||
| L29457.1 | - | 3 | 142 | |||
| L31395.1 | - | 19 | 2013 | |||
| L31396.1 | - | 14 | 1285 | |||
| L31397.1 | - | 18 | 2057 | |||

Dnm1
Genomic Map