siRNA Details
| Gene Symbol: Trdmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010067.4 | NP_034197.3 | 8 | 757 | ||
| GenBank mRNA | AF012129.1 | - | 8 | 744 | ||
| AF045889.1 | - | 8 | 749 | |||
| AK029136.1 | - | 8 | 758 | |||
| AK050403.1 | - | 8 | 757 | |||
| BC087892.1 | - | 8 | 757 | 30504522 | ||

Trdmt1
Genomic Map