siRNA Details
| Gene Symbol: Dok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291799.1 | NP_001278728.1 | 5 | 1124 | ||
| NM_010070.4 | NP_034200.4 | 5 | 1126 | |||
| GenBank mRNA | AK136382.1 | - | 5 | 1127 | ||
| AK169871.1 | - | 4 | 1292 | |||
| BC013066.1 | - | 5 | 1120 | 3498170 | ||
| U78818.2 | - | 5 | 1091 | |||

Dok1
Genomic Map