siRNA Details
| Gene Symbol: Dpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136060.2 | NP_001129532.1 | 3 | 631 | ||
| NM_001198886.1 | NP_001185815.1 | 3 | 715 | |||
| NM_010075.2 | NP_034205.1 | 3 | 966 | |||
| NM_207282.3 | NP_997165.2 | 3 | 819 | |||
| GenBank mRNA | AF092507.1 | - | 3 | 974 | ||
| AK031903.1 | - | 3 | 592 | |||
| AK051708.1 | - | 3 | 797 | |||
| AK082186.1 | - | 3 | 890 | |||
| AK158964.1 | - | 3 | 820 | |||
| BC048383.1 | - | 3 | 482 | 6306267 | ||
| BC085154.1 | - | 3 | 715 | 6838230 | ||

Dpp6
Genomic Map