siRNA Details
| Gene Symbol: Dr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026106.4 | NP_080382.2 | 1 | 745 | ||
| GenBank mRNA | AK031057.1 | - | 1 | 679 | ||
| AK075713.1 | - | 1 | 745 | |||
| AK090113.1 | - | 1 | 727 | |||
| AK139866.1 | - | 1 | 675 | |||
| AK144749.1 | - | 1 | 739 | |||
| AK146573.1 | - | 1 | 708 | |||
| AK217768.1 | - | 1 | 161 | |||
| BC013461.1 | - | 1 | 314 | 4222118 | ||

Dr1
Genomic Map