siRNA Details
| Gene Symbol: Slc26a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021353.3 | NP_067328.1 | 14, 15 | 1771 | ||
| GenBank mRNA | AF136751.1 | - | 14, 15 | 1689 | ||
| BC139271.1 | - | 13, 14 | 1606 | 8862293 | ||
| BC139273.1 | - | 13, 14 | 1606 | 8862295 | ||

Slc26a3
Genomic Map