siRNA Details
| Gene Symbol: Drg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007879.1 | NP_031905.1 | 5 | 590 | ||
| GenBank mRNA | AK044194.1 | - | 5 | 599 | ||
| AK080814.1 | - | 5 | 588 | |||
| AK144438.1 | - | 5 | 592 | |||
| AK155061.1 | - | 5 | 605 | |||
| BC018596.1 | - | 1 | 20 | |||
| BC039649.1 | - | 5 | 584 | 5351885 | ||
| D10715.1 | - | 5 | 590 | |||

Drg1
Genomic Map