siRNA Details
| Gene Symbol: Arid3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288625.1 | NP_001275554.1 | 8 | 1886 | ||
| NM_001288626.1 | NP_001275555.1 | 8 | 2034 | |||
| NM_007880.4 | NP_031906.1 | 8 | 2040 | |||
| GenBank mRNA | AK141283.1 | - | 8 | 1887 | ||
| AK154956.1 | - | 8 | 2012 | |||
| AK155569.1 | - | 6 | 895 | |||
| BC050925.1 | - | 8 | 1938 | 6831312 | ||
| U60335.1 | - | 7 | 1828 | |||

Arid3a
Genomic Map