siRNA Details
| Gene Symbol: Drp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010078.3 | NP_034208.2 | 6 | 968 | ||
| GenBank mRNA | AK034383.1 | - | 6 | 911 | ||
| AK081426.1 | - | 6 | 912 | |||
| AK158102.1 | - | 5 | 770 | |||
| BC060605.1 | - | 5 | 739 | |||
| BC125345.1 | - | 6 | 828 | 40129860 | ||
| BC125347.1 | - | 6 | 828 | 40129862 | ||

Drp2
Genomic Map