siRNA Details
| Gene Symbol: Dsg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007883.2 | NP_031909.2 | 7 | 958 | ||
| GenBank mRNA | AB072269.1 | - | 7 | 986 | ||
| BC020144.1 | - | 7 | 852 | |||
| BC044848.1 | - | 7 | 919 | |||
| BC131653.1 | - | 7 | 934 | 40110529 | ||
| BC131654.1 | - | 7 | 934 | 40110531 | ||

Dsg2
Genomic Map