siRNA Details
| Gene Symbol: Epyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007884.2 | NP_031910.1 | 8 | 994 | ||
| GenBank mRNA | AK005684.1 | - | 8 | 996 | ||
| AK015223.1 | - | 8 | 994 | |||
| AK137179.1 | - | 7 | 919 | |||
| AK157900.1 | - | 1245 | ||||
| AK157901.1 | - | 7 | 1256 | |||
| BC012695.1 | - | 7 | 966 | 4037028 | ||
| D78274.1 | - | 7 | 909 | |||

Epyc
Genomic Map