siRNA Details
| Gene Symbol: Epyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007884.2 | NP_031910.1 | 7 | 852 | ||
| GenBank mRNA | AK005684.1 | - | 7 | 853 | ||
| AK015223.1 | - | 7 | 852 | |||
| AK137179.1 | - | 6 | 777 | |||
| AK157901.1 | - | 6 | 1114 | |||
| BC012695.1 | - | 6 | 824 | 4037028 | ||
| D78274.1 | - | 6 | 767 | |||

Epyc
Genomic Map