siRNA Details
| Gene Symbol: Dtnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162465.1 | NP_001155937.1 | 7 | 955 | ||
| NM_007886.2 | NP_031912.2 | 7 | 890 | |||
| GenBank mRNA | AJ003007.1 | - | 7 | 823 | ||
| AK043436.1 | - | 7 | 913 | |||
| AK083752.1 | - | 7 | 911 | |||
| AK085675.1 | - | 7 | 881 | |||
| AK157158.1 | - | 8 | 912 | |||
| BC030924.1 | - | 7 | 890 | 4910166 | ||
| Y15742.1 | - | 7 | 787 | |||

Dtnb
Genomic Map