siRNA Details
| Gene Symbol: Usp17la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007887.2 | NP_031913.1 | 3 | 1451 | ||
| GenBank mRNA | AY148453.1 | - | 2 | 1280 | ||
| BC140988.1 | - | 3 | 1480 | 9055600 | ||
| U41636.1 | - | 3 | 1463 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp17la| Gene Symbol: Usp17la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007887.2 | NP_031913.1 | 3 | 1451 | ||
| GenBank mRNA | AY148453.1 | - | 2 | 1280 | ||
| BC140988.1 | - | 3 | 1480 | 9055600 | ||
| U41636.1 | - | 3 | 1463 | |||