siRNA Details
| Gene Symbol: Dusp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010090.2 | NP_034220.2 | 3 | 692 | ||
| GenBank mRNA | AK134067.1 | - | 3 | 693 | ||
| BC048696.1 | - | 3 | 678 | 6511513 | ||
| L11330.1 | - | 3 | 647 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp2| Gene Symbol: Dusp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010090.2 | NP_034220.2 | 3 | 692 | ||
| GenBank mRNA | AK134067.1 | - | 3 | 693 | ||
| BC048696.1 | - | 3 | 678 | 6511513 | ||
| L11330.1 | - | 3 | 647 | |||