siRNA Details
| Gene Symbol: Dvl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK155349.1 | - | 4 | 696 | ||
| AK220168.1 | - | 3 | 884 | |||
| BC051991.1 | - | 4 | 455 | |||
| BC088987.1 | - | 4 | 560 | |||
| BC138848.1 | - | 5 | 567 | 8861870 | ||
| BC138849.1 | - | 5 | 567 | 8861871 | ||
| BU702464.1 | - | 4 | 357 | |||
| U10115.1 | - | 4 | 653 | |||

Dvl1
Genomic Map