siRNA Details
| Gene Symbol: Dyrk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113389.1 | NP_001106860.1 | 7 | 1053 | ||
| NM_007890.2 | NP_031916.1 | 7 | 1312 | |||
| GenBank mRNA | AK047845.1 | - | 2 | 300 | ||
| AK052390.1 | - | 1 | 188 | |||
| BC034550.1 | - | 2 | 398 | |||
| BC129888.1 | - | 6 | 915 | 40111469 | ||
| BC129889.1 | - | 6 | 915 | 40111470 | ||
| BC138716.1 | - | 6 | 942 | 8861738 | ||
| BC145405.1 | - | 6 | 915 | 9053963 | ||
| U58497.1 | - | 6 | 942 | |||

Dyrk1a
Genomic Map